1-Heptadecanol ≥97.0%
Leverantör: TCI
Synonyms:
Heptadecan-1-ol, Hexadecyl carbinol, 1-Heptadecanol
H0018-25G
H0018-5G
TCIAH0018-25GEA
2900
SEK
TCIAH0018-25G
TCIAH0018-5G
1-Heptadecanol ≥97.0%
1-Heptadecanol
Formel:
H₃C(CH₂)₁₅CH₂OH Kokpunkt: 308,5 °C (1013 hPa) Smältpunkt: 53…57 °C Densitet: 0,967 g/cm³ (20 °C) Storage Temperature: Omgivande |
MDL Number:
MFCD00002822 CAS nummer: 1454-85-9 EINECS: 215-932-5 |
|
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 53.0 to 56.0 deg-C |
Solubility in hot Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...