Arachidyl alcohol ≥95.0%
Leverantör: TCI EUROPE
Synonyms: 1-Eicosanol
E0004-25G E0004-500G
TCIAE0004-25GEA 285 SEK
TCIAE0004-25G TCIAE0004-500G
Arachidyl alcohol ≥95.0%
Arachidyl alcohol
Formel: H₃C(CH₂)₁₈CH₂OH
MW: 298,55 g/mol
Kokpunkt: 190°C
Smältpunkt: 71,5 °C
Densitet: 0,88 g/cm³ (21 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00002938
CAS nummer: 629-96-9
EINECS: 211-119-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=62240dec5a0eb626785cccc7444df1ae4bdc547e&vid=4e676cafecc3d7b85dad639e7c754728f8552d68&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Melting point 64.0 to 66.0 deg-C
Solubility in hot Methanol very faint turbidity