Nitrometan ≥98.0%

Leverantör: TCI EUROPE
Varning

N0209-25G N0209-500G N0209-100G
TCIAN0209-25GEA 230 SEK
TCIAN0209-25G TCIAN0209-500G TCIAN0209-100G
Nitrometan ≥98.0%
Nitrometan
Formel: CH₃NO₂
MW: 61,04 g/mol
Kokpunkt: 101°C
Smältpunkt: -29°C
Densitet: 1,138 g/cm³ (20 °C)
Flampunkt: 44 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007400
CAS nummer: 75-52-5
EINECS: 200-876-6
UN: 1261
ADR: 3,II
Merck Index: 14,06611
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae4275c74f56c3b61f3218fcff96edf16f955c0c&vid=badad09dce1d23a7f3e6146f5930613703eef1af&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %
agrichemical (Organophosphorus) to pass test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR