1-Ethylpropylamine ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Fara

Synonyms: 3-Pentylamine, 3-Aminopentane, Pentan-3-amine

A1115-25ML A1115-100ML
TCIAA1115-25MLEA 535 SEK
TCIAA1115-25ML TCIAA1115-100ML
1-Ethylpropylamine ≥98.0% (by GC, titration analysis)
1-Ethylpropylamine
Formel: C₅H₁₃N
MW: 87,16 g/mol
Kokpunkt: 91°C
Densitet: 0,7478 g/cm³ (17.5 °C)
Flampunkt: 2°C
MDL Number: MFCD00008096
CAS nummer: 616-24-0
EINECS: 210-471-6
UN: 2733
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=189b3ce54959881c641fd75424614f466dab5d21&vid=189b3ce54959881c641fd75424614f466dab5d21&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR