Propylamin ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
Fara
Synonyms: 1-Aminopropan
P0520-25ML P0520-500ML
TCIAP0520-25MLEA 230 SEK
TCIAP0520-25ML TCIAP0520-500ML
Propylamin ≥98.0% (by GC, titration analysis)
Propylamin
Formel: H₃CCH₂CH₂NH₂
MW: 59,11 g/mol
Kokpunkt: 48°C
Smältpunkt: -83°C
Densitet: 0,7175 g/cm³ (20 °C)
Flampunkt: -30°C
Storage Temperature: Omgivande
MDL Number: MFCD00008205
CAS nummer: 107-10-8
EINECS: 203-462-3
UN: 1277
ADR: 3,II
Merck Index: 14,07843
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9bf4361ef4e08cca4a1009b5680f2da1b0c45aa1&vid=54d11d828139ea09f57fd1ff2189f33b58225e2e&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %