Pentylamine ≥98.0%

Leverantör: TCI EUROPE
Fara

Synonyms: Amylamine, 1-Aminopentane, n-Amylamine, n-Pentylamine, 1-Pentylamine

A0445-25ML A0445-500ML A0445-100ML
TCIAA0445-25MLEA 215 SEK
TCIAA0445-25ML TCIAA0445-500ML TCIAA0445-100ML
Pentylamine ≥98.0%
Pentylamine
Formel: H₃C(CH₂)₃CH₂NH₂
MW: 87,16 g/mol
Kokpunkt: 104°C
Smältpunkt: –50 °C
Densitet: 0,7547 g/cm³ (20 °C)
Flampunkt: -1°C
Storage Temperature: Omgivande
MDL Number: MFCD00008236
CAS nummer: 110-58-7
EINECS: 203-780-2
UN: 1106
ADR: 3,II
Merck Index: 14,00598
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=232c71f0db99ab3d80fe4f5535d29055e7d00b2e&vid=27757a1cfd682356788de031e4d14d34bca87aed&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR