Octylamine ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Fara

Synonyms: 1-Aminooctane, n-Octylamine, 1-Octylamine

O0045-25ML O0045-500ML
TCIAO0045-25MLEA 230 SEK
TCIAO0045-25ML TCIAO0045-500ML
Octylamine ≥98.0% (by GC, titration analysis)
Octylamine
Formel: H₃C(CH₂)₆CH₂NH₂
MW: 129,25 g/mol
Kokpunkt: 176°C
Smältpunkt: –1 °C
Densitet: 0,7812 g/cm³ (20 °C)
Flampunkt: 65°C
Storage Temperature: Omgivande
MDL Number: MFCD00008247
CAS nummer: 111-86-4
EINECS: 203-916-0
UN: 2735
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eb959e0a60a650473d39938f8cf4aad0a335df78&vid=82aefee5f47d10541f4c6b2c0e55a6e62d090775&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR