Octadecylamine ≥85.0%

Leverantör: TCI
Fara

Synonyms: Octadecan-1-amine, 1-Octadecylamine, Stearylamine, 1-Aminooctadecane

O0014-25G O0014-500G
TCIAO0014-25GEA 215 SEK
TCIAO0014-25G TCIAO0014-500G
Octadecylamine ≥85.0%
Octadecylamine
Formel: H₃C(CH₂)₁₆CH₂NH₂
Kokpunkt: 349°C
Smältpunkt: 53°C
Densitet: 0,8618 g/cm³ (20 °C)
Flampunkt: 110 °C
Storage Temperature: Omgivande
MDL Number: MFCD00008159
CAS nummer: 124-30-1
EINECS: 204-695-3
UN: 3077
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=15e189ee24e52670f16ae2d3118eb55dad782377&vid=8392577880f0ccdbefef729ead64251a12a068bc&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 85.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR