Hexylamine ≥99.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
1-Hexylamine, 1-Aminohexane
H0134-25ML
H0134-500ML
H0134-100ML
TCIAH0134-25MLEA
215
SEK
TCIAH0134-25ML
TCIAH0134-500ML
TCIAH0134-100ML
Hexylamine ≥99.0% (by GC, titration analysis)
Hexylamine
Formel:
H₃C(CH₂)₄CH₂NH₂ MW: 101,19 g/mol Kokpunkt: 130°C Smältpunkt: –19 °C Densitet: 0,766 g/cm³ (20 °C) Flampunkt: 27°C Storage Temperature: Omgivande |
MDL Number:
MFCD00008240 CAS nummer: 111-26-2 EINECS: 203-851-8 UN: 2734 ADR: 8,I |
Specifikation testresultat
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 99.0 % |
Purity (Nonaqueous Titration) | min. 99.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...