Hexadecylamine ≥95.0%
Leverantör: TCI
|
Fara
|
Synonyms:
1-Hexadecylamine, n-Hexadecylamine, 1-Aminohexadecane
H0074-25G
H0074-500G
TCIAH0074-25GEA
295
SEK
TCIAH0074-25G
TCIAH0074-500G
Hexadecylamine ≥95.0%
Hexadecylamine
Formel:
H₃C(CH₂)₁₄CH₂NH₂ Kokpunkt: 330°C Smältpunkt: 43…46 °C Densitet: 0,7821 g/cm³ (60 °C) Flampunkt: 140 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00008158 CAS nummer: 143-27-1 EINECS: 205-596-8 UN: 3259 ADR: 8,II |
|
Specifikation testresultat
Appearance | White powder to crystal |
Purity (GC) | min. 95.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...