Dodecylamin ≥97.0% (by GC, titration analysis)
Leverantör: TCI
Fara
D0980-25G D0980-400G
TCIAD0980-25GEA 215 SEK
TCIAD0980-25G TCIAD0980-400G
Dodecylamin ≥97.0% (by GC, titration analysis)
Dodecylamin
Formel: H₃C(CH₂)₁₀CH₂NH₂
Kokpunkt: 259°C
Smältpunkt: 25…28 °C
Densitet: 0,8 g/cm³ (20 °C)
Flampunkt: ≥ 110 °C
Storage Temperature: Omgivande
MDL Number: MFCD00008154
CAS nummer: 124-22-1
EINECS: 204-690-6
UN: 3259
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=146afccfd1d4fef782e78d99863b1d1bfaf7b006&vid=7926c0694e919cffe0113ea18698fbabbe2e3b6a&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %