2-Iodoperfluoropropane ≥95.0%
Leverantör: TCI EUROPE
Varning
Synonyms: 1,1,1,2,3,3,3-Heptafluoro-2-iodopropane, Heptafluoroisopropyl iodide, Heptafluoro-2-iodopropane, 2-Iodoheptafluoropropane
H0629-25G H0629-100G
TCIAH0629-25GEA 645 SEK
TCIAH0629-25G TCIAH0629-100G
2-Iodoperfluoropropane ≥95.0%
2-Iodoperfluoropropane
Formel: C₃F₇I
MW: 295,93 g/mol
Kokpunkt: 40°C
Smältpunkt: –56 °C
Densitet: 2,077 g/cm³ (20 °C)
Storage Temperature: Kylskåp
MDL Number: MFCD00042089
CAS nummer: 677-69-0
EINECS: 211-643-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=31fd82682644cff33a57eebff53422067cab3c14&vid=47daeee2382a8d732d36441da9975c3d066aab56&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow to Light red clear liquid
Purity (GC) min. 95.0 %