1,1,1,2,2-Pentafluoro-4-iodobutane ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
1-Iodo-3,3,4,4,4-pentafluorobutane
P1817-25G
P1817-5G
TCIAP1817-25GEA
3675
SEK
TCIAP1817-25G
TCIAP1817-5G
1,1,1,2,2-Pentafluoro-4-iodobutane ≥98.0%
1,1,1,2,2-Pentafluoro-4-iodobutane
Formel:
C₄H₄F₅I MW: 273,97 g/mol Kokpunkt: 100°C |
MDL Number:
MFCD00155910 CAS nummer: 40723-80-6 EINECS: 255-055-5 ADR: 6.1,III |
Specifikation testresultat
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...