1,1,1,5,5,5-Hexafluoroacetylacetone ≥95.0%

Leverantör: TCI EUROPE
Fara

Synonyms: Hexafluoroacetylacetone, 1,1,1,5,5,5-Hexafluoropentane-2,4-dione, 1,1,1,5,5,5-Hexafluoro-2,4-pentanedione, HFAA

H0476-25G H0476-5G
TCIAH0476-25GEA 1575 SEK
TCIAH0476-25G TCIAH0476-5G
1,1,1,5,5,5-Hexafluoroacetylacetone ≥95.0%
1,1,1,5,5,5-Hexafluoroacetylacetone
Formel: C₅H₂F₆O₂
MW: 208,06 g/mol
Kokpunkt: 70°C
Densitet: 1,47 g/cm³ (20 °C)
Flampunkt: 32°C
Storage Temperature: Omgivande
MDL Number: MFCD00000426
CAS nummer: 1522-22-1
EINECS: 216-191-0
UN: 2920
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6aa9bb4da2687196a00431957fbacc1ee392e6c2&vid=641c8447ba8822bc7cab3d2220bafbd5f5b26a4e&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR