1-Klorpropan ≥99.0%

Leverantör: TCI EUROPE
Fara

C0266-25ML C0266-500ML
TCIAC0266-25MLEA 215 SEK
TCIAC0266-25ML TCIAC0266-500ML
1-Klorpropan ≥99.0%
1-Klorpropan
Formel: C₃H₇Cl
MW: 78,54 g/mol
Kokpunkt: 46°C
Smältpunkt: –122,8 °C
Densitet: 1,3914 g/cm³ (20 °C)
Flampunkt: -31 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00000995
CAS nummer: 540-54-5
EINECS: 208-749-7
UN: 1278
ADR: 3,II
Merck Index: 14,07847
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2c7cc02518aa050be259d251ff4c8695727f8af9&vid=13117ccf0d1e0a728caa4bfcd9a58cf15615688c&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless clear liquid
Purity (GC) min. 99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR