1-Chloroheptane ≥99.0%

Leverantör: TCI
Varning

Synonyms: Heptyl chloride

C0182-25ML C0182-500ML
TCIAC0182-25MLEA 285 SEK
TCIAC0182-25ML TCIAC0182-500ML
1-Chloroheptane ≥99.0%
1-Chloroheptane
Formel: C₇H₁₅Cl
Kokpunkt: 159°C
Smältpunkt: –4 °C
Densitet: 0,877 g/cm³ (20 °C)
Flampunkt: 45°C
Storage Temperature: Omgivande
MDL Number: MFCD00001021
CAS nummer: 629-06-1
EINECS: 211-070-9
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c972857ce7e89598de2bf6aae0ea22d6a80d0119&vid=809e34b2ce7fcce9ef5966df39e17e079665500d&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR