1-Chlorodecane ≥97.0%

Leverantör: TCI
Varning

C0600-25ML C0600-500ML
TCIAC0600-25MLEA 325 SEK
TCIAC0600-25ML TCIAC0600-500ML
1-Chlorodecane ≥97.0%
1-Chlorodecane
Formel: C₁₀H₂₁Cl
Kokpunkt: 223°C
Smältpunkt: –34 °C
Densitet: 0,868 g/cm³ (20 °C)
Flampunkt: 102°C
Storage Temperature: Omgivande
MDL Number: MFCD00000956
CAS nummer: 1002-69-3
EINECS: 213-691-0
UN: 3082
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=486fa37e05dd182f0a0c4f970e08c172551dbbde&vid=45e6e4120881fa326ac1719435e6bcc6d43ccabc&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR