1,6-Dichlorohexane 98,0 GC_ANALYSMETOD

Leverantör: TCI
Varning

D0372-500G D0372-25G
TCIAD0372-500GEA 870 SEK
TCIAD0372-500G TCIAD0372-25G
1,6-Dichlorohexane 98,0 GC_ANALYSMETOD
1,6-Dichlorohexane
Formel: ClH₂C(CH₂)₄CH₂Cl
MW: 155.07 g/mol
Kokpunkt: 95°C
Smältpunkt: -13
Flampunkt: 74°C
Storage Temperature: Omgivande
MDL Number: MFCD00001020
CAS nummer: 2163-00-0
EINECS: 218-491-7
UN: 3082
ADR: 9 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4068ac339d137d085040bbe5de524771158cc9aa&vid=965aaf42867d814a7507fd56ba6dae05c36eced6&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR