1,1,2,2-Tetrakloretan ≥97.0%

Leverantör: TCI EUROPE
Fara

T0063-25G T0063-500G
TCIAT0063-25GEA 245 SEK
TCIAT0063-25G TCIAT0063-500G
1,1,2,2-Tetrakloretan ≥97.0%
1,1,2,2-Tetrakloretan
Formel: Cl₂HCCHCl₂
MW: 167,85 g/mol
Kokpunkt: 146°C
Smältpunkt: -44°C
Densitet: 1,595 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00000848
CAS nummer: 79-34-5
EINECS: 201-197-8
UN: 1702
ADR: 6.1,II
Merck Index: 14,09189
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f145031abedf97ad5c66e1ad218cc0f7459f9b2d&vid=a8fd02f4f04dfb2e7f5c0352998482a7ad98f01b&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR