Tricosanoic acid ≥95.0% (by GC, titration analysis)
Leverantör: TCI
Varning
T0402-1G T0402-10G
TCIAT0402-1GEA 1025 SEK
TCIAT0402-1G TCIAT0402-10G
Tricosanoic acid ≥95.0% (by GC, titration analysis)
Tricosanoic acid
Formel: C₂₃H₄₆O₂
MW: 354.62 g/mol
Kokpunkt: 399
Smältpunkt: 76
Storage Temperature: Kylskåp
MDL Number: MFCD00002808
CAS nummer: 2433-96-7
EINECS: 219-419-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8e014916fbf1b5af803956db0e084bd95a444815&vid=8fb641c38c990f35e8fa180ff0a1734870606891&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Light yellow powder to crystal
Purity (GC) min. 95.0 %
Purity (Neutralization titration) min. 95.0 %
Melting point 78.0 to 81.0 deg-C
Solubility in hot EtOH almost transparency