Myristinsyra ≥99.0%
Leverantör: TCI EUROPE
Varning
Synonyms: Tetradecylic acid, Tetradecanoic acid
M0476-25G M0476-500G
TCIAM0476-25GEA 215 SEK
TCIAM0476-25G TCIAM0476-500G
Myristinsyra ≥99.0%
Myristinsyra
Formel: C₁₄H₂₈O₂
MW: 228,38 g/mol
Kokpunkt: 319 °C (1013 hPa)
Smältpunkt: 51…54 °C
Densitet: 0,8622 g/cm³ (20 °C)
Flampunkt: ≥ 165 °C
Storage Temperature: Frys
MDL Number: MFCD00002744
CAS nummer: 544-63-8
EINECS: 208-875-2
Merck Index: 14,06333
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cee7a7ce7abc7eee84c9034eac608e095fc8f3cb&vid=90c0929a17b60cf7a8c437035069905a786d1abe&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 99.0 %
Melting point 54.0 to 57.0 deg-C
Solubility in Methanol almost transparency