Valerianasyra ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
Fara
Synonyms: Pentansyra
V0003-25ML V0003-500ML
TCIAV0003-25MLEA 230 SEK
TCIAV0003-25ML TCIAV0003-500ML
Valerianasyra ≥98.0% (by GC, titration analysis)
Valerianasyra
Formel: C₅H₁₀O₂
MW: 102,13 g/mol
Kokpunkt: 186°C
Smältpunkt: –32 °C
Densitet: 1,11 g/cm³ (20 °C)
Flampunkt: 96°C
Storage Temperature: Omgivande
MDL Number: MFCD00004413
CAS nummer: 109-52-4
EINECS: 203-677-2
UN: 3265
ADR: 8,II
Merck Index: 14,09904
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11c5107eb93cde517daee7119af10553509d0e7c&vid=192d1cac2b91a651275593d23de0e89bccfa4d83&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %