Stearinsyra ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: Octadecanoic acid, Octadecylic acid

S0163-25G S0163-500G
TCIAS0163-25GEA 215 SEK
TCIAS0163-25G TCIAS0163-500G
Stearinsyra ≥98.0%
Stearinsyra
Formel: C₁₈H₃₆O₂
MW: 284,48 g/mol
Kokpunkt: 240°C
Smältpunkt: 67 °C
Densitet: 0,94 g/cm³ (20 °C)
Flampunkt: (closed cup)
Storage Temperature: Kylskåp
MDL Number: MFCD00002752
CAS nummer: 57-11-4
EINECS: 200-313-4
Merck Index: 14,08804
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=75ff009b9bba4c6d7c28ab3c541f1dcc4c9739f9&vid=f72448c356a8d236535ad340eb7277dd7cec412b&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White powder to crystal
Purity (GC) min. 95.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 68.0 to 71.0 deg-C
Solubility in hot Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR