Nonansyra ≥98.0%
Leverantör: TCI EUROPE
Fara
Synonyms: Nonanoic acid, Pelargonic Acid
P0952-25ML P0952-100ML P0952-500ML
TCIAP0952-25MLEA 215 SEK
TCIAP0952-25ML TCIAP0952-100ML TCIAP0952-500ML
Nonansyra ≥98.0%
Nonansyra
Formel: C₉H₁₈O₂
MW: 158,24 g/mol
Kokpunkt: 254°C
Smältpunkt: 12 °C
Densitet: 0,9055 g/cm³ (20 °C)
Flampunkt: 140°C
Storage Temperature: Kylskåp
MDL Number: MFCD00004433
CAS nummer: 112-05-0
EINECS: 203-931-2
UN: 3265
ADR: 8,III
Merck Index: 14,07070
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a6fbbe29c4f457eba635949355d5dea107093c63&vid=a6fbbe29c4f457eba635949355d5dea107093c63&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %
Purity (Neutralization titration) min. 98.0 %