Nonadecanoic acid ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: Nonadecylic acid

N0283-10G N0283-25G
TCIAN0283-10GEA 1050 SEK
TCIAN0283-10G TCIAN0283-25G
Nonadecanoic acid ≥98.0%
Nonadecanoic acid
Formel: C₁₉H₃₈O₂
MW: 298,51 g/mol
Kokpunkt: 238°C
Smältpunkt: 68…70 °C
Densitet: 0,8468 g/cm³ (70 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00002754
CAS nummer: 646-30-0
EINECS: 211-472-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=62567ede44c280f919206bbec13997789c3218fb&vid=9a8b3401f05935c4e64f68c1e3eecd834e3ad860&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White powder to crystal
Purity (GC) min. 98.0 %
Melting point 67.0 to 70.0 deg-C
Solubility in hot Methanol almost transparency
Neutralization Value 184 to 190

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR