n-Kapronsyra ≥98.0% (by GC, titration analysis)
Leverantör: TCI
Fara
Synonyms: Hexanoic acid
H0105-25ML H0105-500ML
TCIAH0105-25MLEA 245 SEK
TCIAH0105-25ML TCIAH0105-500ML
n-Kapronsyra ≥98.0% (by GC, titration analysis)
n-Kapronsyra
Formel: C₆H₁₂O₂
MW: 116.16 g/mol
Kokpunkt: 205°C
Smältpunkt: –3°C
Flampunkt: 108°C
Storage Temperature: Omgivande
MDL Number: MFCD00004421
CAS nummer: 142-62-1
EINECS: 205-550-7
UN: 2829
ADR: 8 III
Merck Index: 14,01759
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5bbddc56e07d574fb847d5507e056c1b486a81b6&vid=370122cb29e71441bed76a94f46bc986b915280e&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %