Palmitinsyra ≥97.0% (by GC, titration analysis)
Leverantör: TCI
Varning
Synonyms: Hexadecanoic acid, Hexadecylic acid
P0002-25G P0002-500G
TCIAP0002-25GEA 215 SEK
TCIAP0002-25G TCIAP0002-500G
Palmitinsyra ≥97.0% (by GC, titration analysis)
Palmitinsyra
Formel: C₁₆H₃₂O₂
Kokpunkt: 352°C
Smältpunkt: 61…63 °C
Densitet: 0,853 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00002747
CAS nummer: 57-10-3
EINECS: 200-312-9
Merck Index: 14,06996
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=783f66713b15eb34c010030c2b868a6d8625908d&vid=79f5297d5cd66c12a5e4c1711456dac9f968ebb1&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 62.0 to 66.0 deg-C
Solubility in hot Methanol almost transparency