Margaric acid ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
Varning
Synonyms: Heptadecanoic acid, Heptadecylic acid
H0019-25G H0019-500G H0019-100G
TCIAH0019-25GEA 950 SEK
TCIAH0019-25G TCIAH0019-500G TCIAH0019-100G
Margaric acid ≥98.0% (by GC, titration analysis)
Margaric acid
Formel: C₁₇H₃₄O₂
MW: 270,46 g/mol
Kokpunkt: 220°C
Smältpunkt: 63 °C
Densitet: 1,02 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00002751
CAS nummer: 506-12-7
EINECS: 208-027-1
Merck Index: 14,05751
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d9a7ed9d517e3934bd5d8bc729ea44912cfbb41d&vid=b53d67b952755420f33b8d00ba1befd114ed0025&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 59.0 to 63.0 deg-C
Solubility in hot Methanol almost transparency