Isopentylacetat ≥98.0%
Leverantör: TCI EUROPE
Fara
Synonyms: Isopentyl acetate, Acetic acid isoamyl ester
A0033-25ML A0033-500ML
TCIAA0033-25MLEA 230 SEK
TCIAA0033-25ML TCIAA0033-500ML
Isopentylacetat ≥98.0%
Isopentylacetat
Formel: C₇H₁₄O₂
MW: 130,19 g/mol
Kokpunkt: 141°C
Smältpunkt: –78 °C
Densitet: 0,87 g/cm³ (20 °C)
Flampunkt: 38°C
Storage Temperature: Omgivande
MDL Number: MFCD00008946
CAS nummer: 123-92-2
EINECS: 204-662-3
UN: 1104
ADR: 3,III
Merck Index: 14,05111
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4f6b7448766aec298061b4cf89d8531a98e4e6f2&vid=7d254f0205bccf460f477046c624188020b4c16e&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless clear liquid
Purity (GC) min. 98.0 %