Diacetyl ≥98.0%
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
2,3-Butanedione
B0682-25ML
B0682-500ML
B0682-100ML
TCIAB0682-25MLEA
230
SEK
TCIAB0682-25ML
TCIAB0682-500ML
TCIAB0682-100ML
Diacetyl ≥98.0%
Diacetyl
Formel:
C₄H₆O₂ MW: 86,09 g/mol Kokpunkt: 88°C Smältpunkt: –2 °C Densitet: 0,9904 g/cm³ (20 °C) Flampunkt: 11°C Storage Temperature: Kylskåp |
MDL Number:
MFCD00008756 CAS nummer: 431-03-8 EINECS: 207-069-8 UN: 2346 ADR: 3,II Merck Index: 14,02966 |
Specifikation testresultat
Appearance | Light yellow to Yellow to Orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...