3-Mercapto-2-pentanone ≥97.0% (by GC, titration analysis)

Leverantör: TCI EUROPE

M2026-25G M2026-5G
TCIAM2026-25GEA 2500 SEK
TCIAM2026-25G TCIAM2026-5G
3-Mercapto-2-pentanone ≥97.0% (by GC, titration analysis)
3-Mercapto-2-pentanone
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2a0b699f637f83fc0a5a05ccee35fa13742e9d4&vid=1467fe9c0de1b9658f94c005ec3aa0051d28926b&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %
Purity (Iodometric Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR