Hydroxyacetone ≥90.0%

Leverantör: TCI EUROPE
Varning

H0388-25ML H0388-500ML H0388-100ML
TCIAH0388-25MLEA 270 SEK
TCIAH0388-25ML TCIAH0388-500ML TCIAH0388-100ML
Hydroxyacetone ≥90.0%
Hydroxyacetone
Formel: C₃H₆O₂
MW: 74,08 g/mol
Kokpunkt: 145 °C (1013 hPa)
Smältpunkt: –17 °C
Densitet: 1,0928 g/cm³ (20 °C)
Flampunkt: 57°C
Storage Temperature: Kylskåp
MDL Number: MFCD00004669
CAS nummer: 116-09-6
EINECS: 204-124-8
UN: 1993
ADR: 3,III
Merck Index: 13,00066
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2cbdc24c879f6e74de1d223c6d3ea44e629ebb55&vid=3f67ab3ed7fb4a8328b16d53d2a92edb6aa94054&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 90.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR