4-Hydroxybutanone ≥95.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 4-Hydroxy-2-butanone, 4-Hydroxybutan-2-one

H0584-25ML H0584-500ML H0584-100ML
TCIAH0584-25MLEA 565 SEK
TCIAH0584-25ML TCIAH0584-500ML TCIAH0584-100ML
4-Hydroxybutanone ≥95.0%
4-Hydroxybutanone
Formel: C₄H₈O₂
MW: 88,11 g/mol
Kokpunkt: 170°C
Flampunkt: 89°C
MDL Number: MFCD00059005
CAS nummer: 590-90-9
EINECS: 209-693-6
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=24618134bb75112a51ce0d0f8b3860df9009f063&vid=33a48598f5f0dffa030750cb1854e31c4cac18e1&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR