Methoxyacetone ≥96.0%
Leverantör: TCI
|
Varning
|
M0906-5ML
M0906-1ML
TCIAM0906-5MLEA
910
SEK
TCIAM0906-5ML
TCIAM0906-1ML
Methoxyacetone ≥96.0%
Methoxyacetone
Formel:
C₄H₈O₂ Kokpunkt: 115°C Smältpunkt: –66 °C Densitet: 0,957 g/cm³ (20 °C) Flampunkt: 25°C Storage Temperature: Omgivande |
MDL Number:
MFCD00008768 CAS nummer: 5878-19-3 EINECS: 227-549-0 UN: 1993 ADR: 3,III |
|
Specifikation testresultat
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...