9-Heptadecanone ≥98.0%

Leverantör: TCI EUROPE

H0536-25G H0536-5G
TCIAH0536-25GEA 6350 SEK
TCIAH0536-25G TCIAH0536-5G
9-Heptadecanone ≥98.0%
9-Heptadecanone
Formel: C₁₇H₃₄O
MW: 254,46 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00009573
CAS nummer: 540-08-9
EINECS: 208-734-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e49b7fa5476e6c9138c4878f1945b993770b35d7&vid=0b3a34676f2c359de088396df39c2fc6e3427db0&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 50.0 to 54.0 deg-C
Solubility in Toluene almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR