3-tert-Butyltoluene ≥98.0%

Leverantör: TCI EUROPE
Fara

B0967-1G B0967-5G
TCIAB0967-1GEA 1925 SEK
TCIAB0967-1G TCIAB0967-5G
3-tert-Butyltoluene ≥98.0%
3-tert-Butyltoluene
Formel: C₁₁H₁₆
MW: 148,25 g/mol
Kokpunkt: 189°C
Smältpunkt: –41°C
MDL Number: MFCD00059210
CAS nummer: 1075-38-3
UN: 2667
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=02b8f47feb94676af6042da4736367261dead0ce&vid=02b8f47feb94676af6042da4736367261dead0ce&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR