Carbazole ≥97.0%
Leverantör: TCI EUROPE
|
Varning
|
C0032-25G
C0032-500G
C0032-100G
TCIAC0032-25GEA
325
SEK
TCIAC0032-25G
TCIAC0032-500G
TCIAC0032-100G
Carbazole ≥97.0%
Carbazole
Formel:
C₁₂H₉N MW: 167,21 g/mol Kokpunkt: 355 °C (1013 hPa) Smältpunkt: 236 °C Densitet: 1,26 g/cm³ (20 °C) |
MDL Number:
MFCD00004960 CAS nummer: 86-74-8 EINECS: 201-696-0 UN: 3077 ADR: 9,III Merck Index: 14,01790 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Solubility in hot EtOH | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...