4-Propylbenzenesulphonyl chloride ≥93.0%

Leverantör: TCI EUROPE
Fara

Synonyms: 4-Propylbenzene-1-sulphonyl chloride

P1366-25G P1366-5G
TCIAP1366-25GEA 1800 SEK
TCIAP1366-25G TCIAP1366-5G
4-Propylbenzenesulphonyl chloride ≥93.0%
4-Propylbenzenesulphonyl chloride
Formel: C₉H₁₁ClO₂S
MW: 218,7 g/mol
Kokpunkt: 113 - 113,5 °C (0.2 -rr)
Smältpunkt: –4 °C
MDL Number: MFCD00041508
CAS nummer: 146949-07-7
UN: 3265
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1484af68a3e3d573f1f3b8e183c4ddcc37ddeff2&vid=1484af68a3e3d573f1f3b8e183c4ddcc37ddeff2&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to lump
Purity (GC) min. 93.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR