1,1,1-Tris(hydroxymethyl)propane 98,0 GC_ANALYSMETOD
Leverantör: TCI
Synonyms: 1,1,1-Trimethylolpropane, Propylidynetrimethanol, Trimethylol propane, Propylidynetris(methanol), 2-Ethyl-2-(hydroxymethyl)-1,3-propanediol
T0480-25G T0480-500G
TCIAT0480-25GEA 215 SEK
TCIAT0480-25G TCIAT0480-500G
1,1,1-Tris(hydroxymethyl)propane 98,0 GC_ANALYSMETOD
1,1,1-Tris(hydroxymethyl)propane
Formel: C₆H₁₄O₃
MW: 134.18 g/mol
Kokpunkt: 149°C
Smältpunkt: 53
Storage Temperature: Omgivande
MDL Number: MFCD00004694
CAS nummer: 77-99-6
EINECS: 201-074-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3413a080cbfe27f89b3efaf9622596c6b4250232&vid=3413a080cbfe27f89b3efaf9622596c6b4250232&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 58.0 to 62.0 deg-C
Solubility in Water very faint turbidity