4-Hydroxycarbazole ≥98.0%

Leverantör: TCI
Varning

Synonyms: 9H-Carbazol-4-ol, Carbazol-2-ol

H1014-1G H1014-25G H1014-5G
TCIAH1014-1GEA 215 SEK
TCIAH1014-1G TCIAH1014-25G TCIAH1014-5G
4-Hydroxycarbazole ≥98.0%
4-Hydroxycarbazole
Formel: C₁₂H₉NO
MW: 183.21 g/mol
MDL Number: MFCD02178385
CAS nummer: 52602-39-8
EINECS: 258-034-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=105678ae14c4b258549f801ccd2737feccfdb316&vid=105678ae14c4b258549f801ccd2737feccfdb316&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR