3-Phenyl-2-propyn-1-ol ≥97.0%
Leverantör: TCI EUROPE
Synonyms:
3-Phenylprop-2-yn-1-ol
P1147-25G
P1147-5G
TCIAP1147-25GEA
2875
SEK
TCIAP1147-25G
TCIAP1147-5G
3-Phenyl-2-propyn-1-ol ≥97.0%
3-Phenyl-2-propyn-1-ol
Formel:
C₉H₈O MW: 132,16 g/mol Kokpunkt: 145°C |
MDL Number:
MFCD00040914 CAS nummer: 1504-58-1 |
Specifikation testresultat
Appearance | Colorless to Red to Green clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...