(±)-2,5-Dimethyl-3-hexanol ≥97.0%
Leverantör: TCI EUROPE
|
Varning
|
D1969-25ML
D1969-5ML
TCIAD1969-25MLEA
1500
SEK
TCIAD1969-25ML
TCIAD1969-5ML
(±)-2,5-Dimethyl-3-hexanol ≥97.0%
(±)-2,5-Dimethyl-3-hexanol
Formel:
C₈H₁₈O MW: 130,23 g/mol Kokpunkt: 65°C Smältpunkt: –78 °C Densitet: 0,8159 g/cm³ (20 °C) |
MDL Number:
MFCD00026452 CAS nummer: 19550-07-3 EINECS: 243-152-5 UN: 1987 ADR: 3,III |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...