Trimethyl orthobenzoate ≥95.0%

Leverantör: TCI EUROPE

Synonyms: Trimethyl orthobeznoate, (Trimethoxymethyl)benzene

T2287-25G T2287-5G
TCIAT2287-25GEA 1025 SEK
TCIAT2287-25G TCIAT2287-5G
Trimethyl orthobenzoate ≥95.0%
Trimethyl orthobenzoate
Formel: C₁₀H₁₄O₃
MW: 182,22 g/mol
Kokpunkt: 115°C
Flampunkt: 82°C
MDL Number: MFCD00008474
CAS nummer: 707-07-3
EINECS: 211-897-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=68e463574c61de7fb49d27f3340bddfa9ef2f9cd&vid=68e463574c61de7fb49d27f3340bddfa9ef2f9cd&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR