1,6-Diaminohexane dihydrochloride ≥98.0%

Leverantör: TCI
Varning

Synonyms: 1,6-Hexanediamine dihydrochloride, Hexane-1,6-diamine dihydrochloride, Hexamethylenediamine dihydrochloride

D0096-25G D0096-500G
TCIAD0096-25GEA 295 SEK
TCIAD0096-25G TCIAD0096-500G
1,6-Diaminohexane dihydrochloride ≥98.0%
1,6-Diaminohexane dihydrochloride
Formel: C₆H₁₈Cl₂N₂
MW: 189.13 g/mol
Smältpunkt: 258
MDL Number: MFCD00012528
CAS nummer: 6055-52-3
EINECS: 227-977-8
Merck Index: 14,04695
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=77f6dd95011439b0a23253335ea7272af6956824&vid=77f6dd95011439b0a23253335ea7272af6956824&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Solubility in Water very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR