Ethylenediamine monohydrate ≥98.0%
Leverantör: TCI EUROPE
Fara
Synonyms: 1,2-Diaminoethane monohydrate, 1,2-Ethanediamine monohydrate
E0081-25ML E0081-500ML
TCIAE0081-25MLEA 285 SEK
TCIAE0081-25ML TCIAE0081-500ML
Ethylenediamine monohydrate ≥98.0%
Ethylenediamine monohydrate
Formel: C₂H₈N₂·1H₂O
MW: 78,11 g/mol
Kokpunkt: 118°C
Smältpunkt: 10°C
Densitet: 0,926 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00149563
CAS nummer: 6780-13-8
EINECS: 203-468-6
UN: 1604
ADR: 8,II
Merck Index: 14,03795
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=89aa80e70251c02a34b7fca48373af16517f8eef&vid=89aa80e70251c02a34b7fca48373af16517f8eef&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (Neutralization titration) min. 98.0 % (calcd.on anh.substance)
Water 22.0 to 24.0 %