trans-4-Aminocyclohexanol ≥98.0%

Leverantör: TCI
Fara

A1160-25G A1160-500G
TCIAA1160-25GEA 405 SEK
TCIAA1160-25G TCIAA1160-500G
trans-4-Aminocyclohexanol ≥98.0%
trans-4-Aminocyclohexanol
Formel: C₆H₁₃NO
Kokpunkt: 127°C
Smältpunkt: 108…113 °C
MDL Number: MFCD00067698
CAS nummer: 27489-62-9
EINECS: 248-492-8
UN: 3259
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2728d1724c07ce52f0bc46c2ab534b3c344ff0d1&vid=2728d1724c07ce52f0bc46c2ab534b3c344ff0d1&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 109.0 to 113.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR