1-(3-Aminopropyl)imidazole ≥97.0%

Leverantör: TCI EUROPE
Fara

Synonyms: N-(3-Aminopropyl)-imidazole, Imidazole-1-propylamine, 1-(3-Aminopropyl)-1H-imidazole

A1185-25G A1185-500G A1185-100G
TCIAA1185-25GEA 390 SEK
TCIAA1185-25G TCIAA1185-500G TCIAA1185-100G
1-(3-Aminopropyl)imidazole ≥97.0%
1-(3-Aminopropyl)imidazole
Formel: C₆H₁₁N₃
MW: 125,17 g/mol
Kokpunkt: 117°C
Flampunkt: 154°C
MDL Number: MFCD00009819
CAS nummer: 5036-48-6
EINECS: 225-730-9
UN: 2735
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=590337d03d4baa8ec42d149a3755daa964547320&vid=590337d03d4baa8ec42d149a3755daa964547320&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR