2-(Trifluoromethyl)phenylacetonitrile ≥98.0%
Leverantör: TCI EUROPE
Fara
Synonyms: 2-(Trifluoromethyl)benzyl cyanide, 1-(Cyanmethyl)-2-(trifluormethyl)benzene
T2366-25G T2366-5G
TCIAT2366-25GEA 1550 SEK
TCIAT2366-25G TCIAT2366-5G
2-(Trifluoromethyl)phenylacetonitrile ≥98.0%
2-(Trifluoromethyl)phenylacetonitrile
Formel: C₉H₆F₃N
MW: 185,15 g/mol
Kokpunkt: 92°C
Smältpunkt: 29…30 °C
MDL Number: MFCD00001903
CAS nummer: 3038-47-9
EINECS: 221-239-9
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=80012c18a2d06d799ec0ca0cf7579f0fc3891fd2&vid=80012c18a2d06d799ec0ca0cf7579f0fc3891fd2&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White or Colorless to Yellow to Orange powder to lump to clear liquid
Purity (GC) min. 98.0 %