2-Fluoro-5-nitrobenzaldehyde ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
F0618-1G
F0618-5G
TCIAF0618-1GEA
565
SEK
TCIAF0618-1G
TCIAF0618-5G
2-Fluoro-5-nitrobenzaldehyde ≥98.0%
2-Fluoro-5-nitrobenzaldehyde
Formel:
C₇H₄FNO₃ MW: 169,11 g/mol |
MDL Number:
MFCD00042298 CAS nummer: 27996-87-8 |
Specifikation testresultat
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 57.0 to 61.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...