Ethoxyacetic acid ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Fara

Synonyms: 2-Ethoxyacetic acid

E0377-25G E0377-500G E0377-100G
TCIAE0377-25GEA 565 SEK
TCIAE0377-25G TCIAE0377-500G TCIAE0377-100G
Ethoxyacetic acid ≥98.0% (by GC, titration analysis)
Ethoxyacetic acid
Formel: C₄H₈O₃
MW: 104,11 g/mol
Kokpunkt: 207°C
Densitet: 1,107 g/cm³ (11 °C)
Flampunkt: 97°C
MDL Number: MFCD00004310
CAS nummer: 627-03-2
EINECS: 210-978-2
UN: 3265
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9f0ed660bdb546b543fa6bc711feb1b42fcf1a5f&vid=9f0ed660bdb546b543fa6bc711feb1b42fcf1a5f&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR